CH4424846606 ChemBridge Corporation

N-methyl-N-phenylglycine hydrochloride

  • MDL number MFCD13186269
  • Cas Number : 21911-75-1
  • Molecular Weight : 201.6501

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4026827

Safety Information

MSDS https://www.hit2lead.com/sds/4026827_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart