CH4424846607 ChemBridge Corporation

3-(3-piperidinyl)propanoic acid hydrochloride

  • MDL number MFCD03453312
  • Cas Number : 71985-82-5
  • Molecular Weight : 193.6711

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4026834

Safety Information

MSDS https://www.hit2lead.com/sds/4026834_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart