CH4424846610 ChemBridge Corporation

2-methyl-2-(4-morpholinyl)propanoic acid hydrochloride hydrate

  • MDL number MFCD26959521
  • Cas Number : 87439-10-9
  • Molecular Weight : 227.6858

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl H2O
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://www.hit2lead.com/building-blocks/4026853

Safety Information

msds https://www.hit2lead.com/building-blocks/4026853
Availability details for
Quantity
  • Availability
  • Add to Cart