CH4424846611 ChemBridge Corporation

4-(1-aminoethyl)phenol hydrochloride

  • MDL number MFCD11226473
  • Cas Number : 860767-47-1
  • Molecular Weight : 173.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4026855

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4026855
Availability details for
Quantity
  • Availability
  • Add to Cart