CH4424846617 ChemBridge Corporation

N-cyclopropylcyclohexanamine hydrochloride

  • MDL number MFCD11696449
  • Cas Number : 874-64-6
  • Molecular Weight : 175.6989

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4026926

Safety Information

MSDS https://www.hit2lead.com/sds/4026926_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart