CH4424846627 ChemBridge Corporation

(5-chloro-2-isopropoxybenzyl)amine hydrochloride

  • MDL number MFCD11226477
  • Cas Number : 1135292-94-2
  • Molecular Weight : 236.1382

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4027006

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4027006
Availability details for
Quantity
  • Availability
  • Add to Cart