CH4424846637 ChemBridge Corporation

(4-isopropoxy-3-methoxybenzyl)amine hydrochloride

  • MDL number MFCD09991467
  • Cas Number : 854185-11-8
  • Molecular Weight : 231.7191

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4027077

Safety Information

MSDS https://www.hit2lead.com/sds/4027077_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart