CH4424846645 ChemBridge Corporation

(3-methoxy-2-propoxybenzyl)amine hydrochloride

  • MDL number MFCD09991466
  • Cas Number : 89411-11-0
  • Molecular Weight : 231.7191

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4027129

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4027129
Availability details for
Quantity
  • Availability
  • Add to Cart