CH4424846660 ChemBridge Corporation

N-(tert-butyl)-2-(1-piperazinyl)acetamide dihydrochloride

  • MDL number MFCD07290110
  • Cas Number : 1044707-29-0
  • Molecular Weight : 272.2151

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4027245

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4027245
Availability details for
Quantity
  • Availability
  • Add to Cart