CH4424846714 ChemBridge Corporation

(3-propoxybenzyl)amine hydrochloride

  • MDL number MFCD10687270
  • Cas Number : 467462-57-3
  • Molecular Weight : 201.6931

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4027733

Safety Information

MSDS https://www.hit2lead.com/sds/4027733_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart