CH4424846732 ChemBridge Corporation

(5-chloro-2-propoxybenzyl)amine hydrochloride

  • MDL number MFCD11226481
  • Cas Number : 1135288-57-1
  • Molecular Weight : 236.1382

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4027916

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4027916
Availability details for
Quantity
  • Availability
  • Add to Cart