CH4424846773 ChemBridge Corporation

[2-(benzyloxy)ethyl]amine hydrochloride

  • MDL number MFCD06411550
  • Cas Number : 10578-75-3
  • Molecular Weight : 187.6666

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4028211

Safety Information

MSDS https://www.hit2lead.com/sds/4028211_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart