CH4424846790 ChemBridge Corporation

1-(3-chloropropyl)-2-methyl-1H-imidazole hydrochloride

  • MDL number MFCD22392161
  • Cas Number : 1421601-58-2
  • Molecular Weight : 195.0896

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4028350

Safety Information

MSDS https://www.hit2lead.com/sds/4028350_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart