CH4424846819 ChemBridge Corporation

N,3,3-trimethyl-2-butanamine hydrochloride

  • MDL number MFCD09971468
  • Cas Number : 89979-68-0
  • Molecular Weight : 151.6775

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4028680

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4028680
Availability details for
Quantity
  • Availability
  • Add to Cart