CH4424846823 ChemBridge Corporation

[1-(4-propoxyphenyl)propyl]amine hydrochloride

  • MDL number MFCD10703566
  • Cas Number : 1201633-55-7
  • Molecular Weight : 229.7463

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4028719

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4028719
Availability details for
Quantity
  • Availability
  • Add to Cart