CH4424846838 ChemBridge Corporation

N~1~-(tert-butyl)glycinamide hydrochloride

  • MDL number MFCD07324741
  • Cas Number : 80008-05-5
  • Molecular Weight : 166.6491

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4028896

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4028896
Availability details for
Quantity
  • Availability
  • Add to Cart