CH4424846890 ChemBridge Corporation

(2-ethoxypropyl)amine hydrochloride

  • MDL number MFCD09997700
  • Cas Number : 1184979-76-7
  • Molecular Weight : 139.6238

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4029821

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4029821
Availability details for
Quantity
  • Availability
  • Add to Cart