CH4424846897 ChemBridge Corporation

1-benzyl-1H-pyrazol-4-amine hydrochloride

  • MDL number MFCD18425906
  • Cas Number : 1264097-17-7
  • Molecular Weight : 209.6754

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4029886

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4029886
Availability details for
Quantity
  • Availability
  • Add to Cart