CH4424846898 ChemBridge Corporation

1,5-dimethyl-1H-pyrazol-4-amine dihydrochloride

  • MDL number MFCD04969122
  • Cas Number : 1189950-55-7
  • Molecular Weight : 184.0669

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4029888

Safety Information

MSDS https://www.hit2lead.com/sds/4029888_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart