CH4424847028 ChemBridge Corporation

trans-3,4-pyrrolidinediyldimethanol hydrochloride

  • MDL number MFCD26959332
  • Cas Number : 1609409-08-6
  • Molecular Weight : 167.6339

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://www.hit2lead.com/building-blocks/4034788

Safety Information

msds https://www.hit2lead.com/building-blocks/4034788
Availability details for
Quantity
  • Availability
  • Add to Cart