CH4424847098 ChemBridge Corporation

3-methyl-5-(2-pyrrolidinyl)-1H-1,2,4-triazole dihydrochloride

  • MDL number MFCD28024766
  • Cas Number : 1609403-06-6
  • Molecular Weight : 225.1189

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045560

Safety Information

MSDS https://www.hit2lead.com/sds/4045560_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart