CH4424847102 ChemBridge Corporation

3-cyclopropyl-5-[(2S)-2-pyrrolidinyl]-1,2,4-oxadiazole hydrochloride

  • MDL number MFCD28024770
  • Cas Number : 1042690-36-7
  • Molecular Weight : 215.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045581

Safety Information

MSDS https://www.hit2lead.com/sds/4045581_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart