CH4424847124 ChemBridge Corporation

1-ethyl-1H-benzimidazol-5-amine dihydrochloride

  • MDL number MFCD09997670
  • Cas Number : 1185293-97-3
  • Molecular Weight : 234.1256

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4031586

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4031586
Availability details for
Quantity
  • Availability
  • Add to Cart