CH4424847145 ChemBridge Corporation

2-(2-furyl)azepane hydrochloride

  • MDL number MFCD18483486
  • Cas Number : 1269053-71-5
  • Molecular Weight : 201.6931

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data HCl
Assay 0.95
Storage temp. Room Temperature

Safety Information

msds https://www.hit2lead.com/building-blocks/4031895
Availability details for
Quantity
  • Availability
  • Add to Cart