CH4424847180 ChemBridge Corporation

[2-(2-propoxyphenyl)ethyl]amine hydrochloride

  • MDL number MFCD12195855
  • Cas Number : 1201633-42-2
  • Molecular Weight : 215.7197

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4032322

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4032322
Availability details for
Quantity
  • Availability
  • Add to Cart