CH4424847214 ChemBridge Corporation

[2-(4-propoxyphenyl)ethyl]amine hydrochloride

  • MDL number MFCD01657175
  • Cas Number : 56370-31-1
  • Molecular Weight : 215.7197

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4032853

Safety Information

MSDS https://www.hit2lead.com/sds/4032853_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart