CH4424847257 ChemBridge Corporation

2-(chloromethyl)-3,5-dimethyl-4(1H)-pyridinone hydrochloride

  • MDL number MFCD12026993
  • Cas Number : 220770-99-0
  • Molecular Weight : 208.085

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4033440

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4033440
Availability details for
Quantity
  • Availability
  • Add to Cart