CH4424847275 ChemBridge Corporation

{4-[2-(2-pyridinyl)ethyl]phenyl}amine dihydrochloride

  • MDL number MFCD06801252
  • Cas Number : 1185296-34-7
  • Molecular Weight : 271.1855

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4033581

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4033581
Availability details for
Quantity
  • Availability
  • Add to Cart