CH4424847285 ChemBridge Corporation

N-(2-aminoethyl)-N-methylmethanesulfonamide hydrochloride

  • MDL number MFCD18205944
  • Cas Number : 569354-11-6
  • Molecular Weight : 188.6762

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4033691

Safety Information

MSDS https://www.hit2lead.com/sds/4033691_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart