CH4424847288 ChemBridge Corporation

N~1~-methylalaninamide hydrochloride

  • MDL number MFCD13562337
  • Cas Number : 38215-73-5
  • Molecular Weight : 138.5959

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4033710

Safety Information

MSDS https://www.hit2lead.com/sds/4033710_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart