CH4424847304 ChemBridge Corporation

{2-[3-(3-methylbutoxy)phenyl]ethyl}amine hydrochloride

  • MDL number MFCD13248799
  • Cas Number : 1201633-60-4
  • Molecular Weight : 243.7729

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4033933

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4033933
Availability details for
Quantity
  • Availability
  • Add to Cart