CH4424847326 ChemBridge Corporation

[cyclopropyl(4-methyl-2-pyridinyl)methyl]amine dihydrochloride

  • MDL number MFCD18205892
  • Cas Number : 1269053-80-6
  • Molecular Weight : 235.1534

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4034248

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4034248
Availability details for
Quantity
  • Availability
  • Add to Cart