CH4424847330 ChemBridge Corporation

2-(1H-imidazol-1-yl)-3-pyridinamine dihydrochloride

  • MDL number MFCD11802533
  • Cas Number : 1171804-01-5
  • Molecular Weight : 233.0978

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data 2HCl
Assay 0.95
Storage temp. Room Temperature

Safety Information

msds https://www.hit2lead.com/building-blocks/4034305
Availability details for
Quantity
  • Availability
  • Add to Cart