CH4424847355 ChemBridge Corporation

1-methyl-1H-imidazol-2-amine hydrochloride

  • MDL number MFCD10697505
  • Cas Number : 1450-94-8
  • Molecular Weight : 133.5794

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4034680

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4034680
Availability details for
Quantity
  • Availability
  • Add to Cart