CH4424847398 ChemBridge Corporation

3-(1-piperazinyl)-1,2-benzisothiazole hydrochloride

  • MDL number MFCD00674131
  • Cas Number : 87691-88-1
  • Molecular Weight : 255.767

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4034980

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4034980
hstatements H302, H319, H317, H361f, H410.
Availability details for
Quantity
  • Availability
  • Add to Cart