CH4424847407 ChemBridge Corporation

[1-(3-ethyl-5-isoxazolyl)ethyl]amine hydrochloride

  • MDL number MFCD26959656
  • Cas Number : 1609403-26-0
  • Molecular Weight : 176.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data HCl
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035144

Safety Information

MSDS https://www.hit2lead.com/sds/4035144_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart