CH4424847450 ChemBridge Corporation

N-methyl-1-(2-naphthyl)ethanamine hydrochloride

  • MDL number MFCD13195885
  • Cas Number : 1197565-94-8
  • Molecular Weight : 221.7258

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035747

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4035747
Availability details for
Quantity
  • Availability
  • Add to Cart