CH4424847476 ChemBridge Corporation

N-methyl-beta-alanine hydrochloride

  • MDL number MFCD03490507
  • Cas Number : 65845-56-9
  • Molecular Weight : 139.5807

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4036102

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4036102
Availability details for
Quantity
  • Availability
  • Add to Cart