CH4424847483 ChemBridge Corporation

4-methyl-1H-benzimidazol-2-amine hydrobromide

  • MDL number MFCD26959414
  • Cas Number : 1609406-56-5
  • Molecular Weight : 228.0891

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4036205

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4036205
Availability details for
Quantity
  • Availability
  • Add to Cart