CH4424847485 ChemBridge Corporation

1-(1,1-dioxidotetrahydro-2H-thiopyran-4-yl)-4-piperidinamine dihydrochloride

  • MDL number MFCD13461777
  • Cas Number : 2103401-19-8
  • Molecular Weight : 305.2649

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4036265

Safety Information

MSDS https://www.hit2lead.com/sds/4036265_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart