CH4424847534 ChemBridge Corporation

spiro[indole-3,4'-piperidin]-2(1H)-one hydrochloride

  • MDL number MFCD15071694
  • Cas Number : 356072-46-3
  • Molecular Weight : 238.7133

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4036843

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4036843
Availability details for
Quantity
  • Availability
  • Add to Cart