CH4424847542 ChemBridge Corporation

3-(3-azetidinyl)pyridine dihydrochloride

  • MDL number MFCD20527356
  • Cas Number : 1236791-61-9
  • Molecular Weight : 207.1003

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4037129

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4037129
Availability details for
Quantity
  • Availability
  • Add to Cart