CH4424847591 ChemBridge Corporation

7-methyl-2,7-diazaspiro[4.5]decan-6-one hydrochloride

  • MDL number MFCD10700086
  • Cas Number : 1203682-44-3
  • Molecular Weight : 204.6971

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4038667

Safety Information

MSDS https://www.hit2lead.com/sds/4038667_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart