CH4424847628 ChemBridge Corporation

1-benzyl-2-(chloromethyl)-1H-imidazole hydrochloride

  • MDL number MFCD00035210
  • Cas Number : 19276-03-0
  • Molecular Weight : 243.1324

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4039853

Safety Information

MSDS https://www.hit2lead.com/sds/4039853_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart