CH4424847645 ChemBridge Corporation

3-methyl-4H-1,2,4-triazol-4-amine hydrochloride

  • MDL number MFCD16039354
  • Cas Number : 56873-82-6
  • Molecular Weight : 134.5675

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040061

Safety Information

MSDS https://www.hit2lead.com/sds/4040061_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart