CH4424847662 ChemBridge Corporation

[3-(benzyloxy)benzyl]amine hydrochloride hydrate

  • MDL number MFCD18483559
  • Molecular Weight : 267.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl H2O
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4040283

Safety Information

MSDS https://www.hit2lead.com/sds/4040283_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart