CH4424847664 ChemBridge Corporation

5-(1-amino-1-methylethyl)-1,3,4-thiadiazol-2-amine trihydrochloride

  • MDL number MFCD26959412
  • Cas Number : 1609403-73-7
  • Molecular Weight : 267.6075

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040306

Safety Information

MSDS https://www.hit2lead.com/sds/4040306_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart