CH4424847680 ChemBridge Corporation

{2-[5-(tetrahydro-2-furanyl)-1,2,4-oxadiazol-3-yl]ethyl}amine hydrochloride

  • MDL number MFCD19103489
  • Cas Number : 1332529-78-8
  • Molecular Weight : 219.6687

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040602

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4040602
Availability details for
Quantity
  • Availability
  • Add to Cart