CH4424847682 ChemBridge Corporation

1,4'-bipiperidine-3-carboxylic acid dihydrochloride

  • MDL number MFCD06799961
  • Cas Number : 1185293-24-6
  • Molecular Weight : 285.2106

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040615

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4040615
Availability details for
Quantity
  • Availability
  • Add to Cart