CH4424847685 ChemBridge Corporation

[2-(5-cyclopentyl-1,2,4-oxadiazol-3-yl)ethyl]amine hydrochloride

  • MDL number MFCD19103490
  • Cas Number : 1244058-78-3
  • Molecular Weight : 217.6958

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040719

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4040719
Availability details for
Quantity
  • Availability
  • Add to Cart